missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Immunogen sequence: TKLDLRDDKD TIERLRDKKL APITYPQGLA MAREI Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
The protein encoded by this gene is a GTPase which belongs to the RAS superfamily of small GTP-binding proteins. Members of this superfamily appear to regulate a diverse array of cellular events, including the control of cell growth, cytoskeletal reorganization, and the activation of protein kinases. Two transcript variants encoding different isoforms have been found for this gene.
Especificaciones
Especificaciones
| Antígeno | RAC1/RAC2/RAC3 |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.1 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| génica | RAC2 |
| N.º de referencia del gen | P15153, P60763, P63000 |
| Alias de gen | AI323801; AI452260; AL023026; Cell migration-inducing gene 5 protein; D5Ertd559e; EN-7; EN-7 protein; GTPase; Gx; HSPC022; MIG5; p21-Rac1; p21-Rac2; p21-Rac3; protein EN-7; rac; Rac family small GTPase 1; Rac family small GTPase 1 L homeolog; Rac family small GTPase 2; Rac family small GTPase 3; rac1; Rac-1; rac1.L; Rac1B; RAC2; Rac3; Ras-like protein TC25; RAS-related C3 botulinum substrate 1; RAS-related C3 botulinum substrate 2; RAS-related C3 botulinum substrate 3; ras-related C3 botulinum toxin substrate 1; ras-related C3 botulinum toxin substrate 1 (rho family small GTP binding protein Rac1); ras-related C3 botulinum toxin substrate 1 (rho family, small GTP binding protein Rac1); ras-related C3 botulinum toxin substrate 1 (rho family, small GTP binding protein Rac1) L homeolog; ras-related C3 botulinum toxin substrate 2; ras-related C3 botulinum toxin substrate 2 (rho family, small GTP binding protein Rac2); ras-related C3 botulinum toxin substrate 3; ras-related C3 botulinum toxin substrate 3 (rho family, small GTP binding protein Rac3); Ras-related C3 botulinum toxin substrate 3 (rho family, small GTP-binding protein Rac2); rho family, small GTP binding protein Rac1; rho family, small GTP binding protein Rac3; small G protein; TC25; TC-25; XELAEV_18045321mg; Xrac |
| Símbolos de los genes | Rac1, RAC2, Rac3 |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?
