missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rac1 Mouse anti-Human, Mouse, Rat, Clone: 4A4B11, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 66122-1-IG-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The protein encoded by this gene is a GTPase which belongs to the RAS superfamily of small GTP-binding proteins. Members of this superfamily appear to regulate a diverse array of cellular events, including the control of cell growth, cytoskeletal reorganization, and the activation of protein kinases. Two transcript variants encoding different isoforms have been found for this gene.Especificaciones
| Rac1 | |
| Monoclonal | |
| 0.7 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P63000, P63001, Q6RUV5 | |
| Rac1 | |
| Rac1 Fusion Protein Ag17779 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 4A4B11 | |
| Unconjugated | |
| RAC1 | |
| MIG5, p21 Rac1, RAC1, RAC1b, Ras like protein TC25, TC 25, TC25 | |
| Mouse | |
| Protein A | |
| RUO | |
| 19353, 363875, 5879 | |
| -20°C | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido