Learn More
rac Dihydro Mebendazole, TRC
Código de producto. 30004188
Change view
Click to view available options
Cantidad:
50 mg
500 mg
Tamaño de la unidad:
50 miligramos
500 miligramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
30004188 50 mg 50 miligramos
30135607 500 mg 500 miligramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Código de producto. 30004188 Proveedor Toronto Research Chemicals N.º de proveedor TRCD44932050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
CAS 60254-95-7
Nombre del producto químico o material Mebendazole-5-hydroxy
Formula Weight (peso de la fórmula) 297.1113
Fórmula InChI InChI=1S/C16H15N3O3/c1-22-16(21)19-15-17-12-8-7-11(9-13(12)18-15)14(20)10-5-3-2-4-6-10/h2-9,14,20H,1H3,(H2,17,18,19,21)
Nombre IUPAC methyl N-[5-[hydroxy(phenyl)methyl]-1H-benzimidazol-2-yl]carbamate
Fórmula molecular C16 H15 N3 O3
Peso molecular (g/mol) 297.31
Porcentaje de pureza >95%
Notas de grado de pureza HPLC
SMILES COC(=O)Nc1nc2cc(ccc2[nH]1)C(O)c3ccccc3
Sinónimo Carbamic acid, [5-(hydroxyphenylmethyl)-1H-benzimidazol-2-yl]-, methyl ester (9CI), R 19167