missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF347, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant ZNF347.
Marca: Abnova PAB21573.100uL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Sequence: LGISCFDLSIISMLEQGKEPFTLESQVQIAGNPDGWEWIKAVITALSSEFVMKDLLHKGKSNTGEVFQTVMEspecificaciones
| ZNF347 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant ZNF347. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| ZNF1111 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 84671 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:200-1:500) The optimal working dilution should be determined by the end user. | |
| ZNF347 | |
| ZNF347 | |
| Recombinant protein corresponding to amino acids of human ZNF347. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction