Learn More
VENTX, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody rasied against synthetic peptide of VENTX.
Marca: Abnova PAB19285.100ug
Descripción
This gene encodes a member of the Vent family of homeodomain proteins. The encoded protein may function as a transcriptional repressor and be involved in mesodermal patterning and hemopoietic stem cell maintenance. Multiple pseudogenes exist for this gene. A transcribed pseudogene located on chromosome X may lead to antigen production in certain melanomas. [provided by RefSeq
Specifications
| VENTX | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) Immunohistochemistry (5 ug/mL) The optimal working dilution should be determined by the end user. | |
| VENTX | |
| VENTX | |
| A synthetic peptide corresponding to 16 amino acids near internal region of human VENTX. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody rasied against synthetic peptide of VENTX. | |
| In PBS (0.02% sodium azide) | |
| HPX42B/MGC119910/MGC119911/NA88A/VENTX2 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 27287 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only