missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TUBB Rabbit anti-Chicken, Human, Mouse, Pig, Primates, Rat, Xenopus, Zebra fish, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TUBB.
Marca: Abnova PAB11989.100uL
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
| TUBB | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of TUBB. | |
| In PBS (0.02% sodium azide) | |
| M40/MGC117247/MGC16435/OK/SW-cl.56/TUBB1/TUBB5 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| This antibody is specific to beta tubulin. | |
| Store at 4°C for short term. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| TUBB | |
| TUBB | |
| A synthetic peptide corresponding to amino acids 1-100 of human TUBB. | |
| RUO | |
| 203068 | |
| Chicken, Human, Mouse, Pig, Primate, Rat, Xenopus species, Zebrafish | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido