Learn More
TRIM29 Rabbit anti-Bovine, Chimpanzee, Horse, Human, Macaque, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against partial recombinant TRIM29.
Marca: Abnova PAB9977.100ug
Descripción
The protein encoded by this gene belongs to the TRIM protein family. It has multiple zinc finger motifs and a leucine zipper motif. It has been proposed to form homo- or heterodimers which are involved in nucleic acid binding. Thus, it may act as a transcriptional regulatory factor involved in carcinogenesis and/or differentiation. It may also function in the suppression of radiosensitivity since it is associated with ataxia telangiectasia phenotype. [provided by RefSeq
Especificaciones
| TRIM29 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against partial recombinant TRIM29. | |
| In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| ATDC/FLJ36085 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody reacts with over-expressed, acetylated lysine ATDC protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:100000) Western Blot (1:500-1:3000) The optimal working dilution should be determined by the end user. | |
| TRIM29 | |
| TRIM29 | |
| Recombinant protein corresponding to internal region around acetylated K116 of human TRIM29. | |
| RUO | |
| 23650 | |
| Bovine, Chimpanzee, Equine, Human, Macaque, Mouse, Rat | |
| Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only