missing translation for 'onlineSavingsMsg'
Learn More

TNNI3 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Código de producto. 16152820
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
Código de producto. Cantidad unitSize
16152820 100 μg 100 microgramos
1 options
This item is not returnable. View return policy

Product Code. 16152820

missing translation for 'mfr': Abnova PAB18306.100ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against synthetic peptide of TNNI3.

Troponin I (TnI), along with troponin T (TnT) and troponin C (TnC), is one of 3 subunits that form the troponin complex of the thin filaments of striated muscle. TnI is the inhibitory subunit; blocking actin-myosin interactions and thereby mediating striated muscle relaxation. The TnI subfamily contains three genes: TnI-skeletal-fast-twitch, TnI-skeletal-slow-twitch, and TnI-cardiac. This gene encodes the TnI-cardiac protein and is exclusively expressed in cardiac muscle tissues. Mutations in this gene cause familial hypertrophic cardiomyopathy type 7 (CMH7) and familial restrictive cardiomyopathy (RCM). [provided by RefSeq

Specifications

Antígeno TNNI3
Aplicaciones ELISA, Immunohistochemistry (PFA fixed), Western Blot
Clasificación Polyclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Descripción Rabbit polyclonal antibody raised against synthetic peptide of TNNI3.
Dilución Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:100) ELISA (1:5000) The optimal working dilution should be determined by the end user.
Formulación In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide)
génica TNNI3
Alias de gen CMD2A/CMH7/MGC116817/RCM1/TNNC1/cTnI
Símbolos de los genes TNNI3
Especie del huésped Rabbit
Inmunógeno A synthetic peptide corresponding to residues surrounding S43 of human TNNI3.
Método de purificación Affinity purification
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 7137
Especificidad de la prueba This antibody is specific to TNNI3.
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Formulario Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.