missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TBC1D23 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant TBC1D23.
Marca: Abnova PAB23230.100uL
Este artículo no se puede devolver.
Vea la política de devoluciones
Description
Sequence: HLSTAFHLDSDLMLQNPSEFAQSVKSLLEAQKQSIESGSIAGGEHLCFMGSGREEEDMYMNMVLAHFLQKNKEYVSIASGGFMALQQHLADINVDGPENGYSpécification
| TBC1D23 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant TBC1D23. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| DKFZp667G062/FLJ11046/NS4ATP1 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 55773 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:10-1:20) The optimal working dilution should be determined by the end user. | |
| TBC1D23 | |
| TBC1D23 | |
| Recombinant protein corresponding to amino acids of human TBC1D23. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu