Learn More
SYT6 (phospho T418), Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of SYT6.
Marca: Abnova PAB17103.100ug
Descripción
Synaptotagmins, such as SYT6, share a common domain structure that includes a transmembrane domain and a cytoplasmic region composed of 2 C2 domains. Some synaptotagmins are involved in synaptic membrane fusion, while others have a more general function in endocytosis. For further information on synaptotagmins, see MIM 185605.[supplied by OMIM
Especificaciones
| SYT6 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of SYT6. | |
| In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide) | |
| SYT6 | |
| Synthetic phosphopeptide corresponding to residues surrounding T418 of human SYT6. | |
| RUO | |
| 148281 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| SYT6 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only