Learn More
SYT1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SYT1.
Marca: Abnova PAB26807.100ug
Descripción
The synaptotagmins are integral membrane proteins of synaptic vesicles thought to serve as Ca(2+) sensors in the process of vesicular trafficking and exocytosis. Calcium binding to synaptotagmin I participates in triggering neurotransmitter release at the synapse (Fernandez-Chacon et al., 2001 [PubMed 11242035]).[supplied by OMIM
Sequence: G-L-Sp-D-PEspecificaciones
| SYT1 | |
| Polyclonal | |
| Unconjugated | |
| Immunofluorescence (1:100-1:200) The optimal working dilution should be determined by the end user. | |
| SYT1 | |
| SYT1 | |
| A synthetic peptide (conjugated with KLH) corresponding to residues surrounding S309 of human SYT1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Immunofluorescence | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of SYT1. | |
| In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
| DKFZp781D2042/P65/SVP65/SYT | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 6857 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only