Learn More
STOX1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of STOX1.
Marca: Abnova PAB27194.100ug
Descripción
The protein encoded by this gene may function as a DNA binding protein. Mutations in this gene are associated with pre-eclampsia/eclampsia 4 (PEE4). Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Especificaciones
| STOX1 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| STOX1 | |
| STOX1 | |
| A synthetic peptide corresponding to 14 amino acids at C-terminus of human STOX1. | |
| 100 μg | |
| Primary | |
| STOX1 antibody is human reactive. At least three alternatively spliced transcript variants encoding distinct isoforms have been observed. STOX1 antibody recognize the longest isoform. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| ELISA, Immunofluorescence, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of STOX1. | |
| In PBS (0.02% sodium azide) | |
| C10orf24/PEE4 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 219736 | |
| Human | |
| Liquid |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
For Research Use Only