missing translation for 'onlineSavingsMsg'
Learn More

SLIT2 Rabbit anti-Bovine, Chicken, Clawed frog, Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Product Code. 16089326
Change view
Click to view available options
Cantidad:
100 μg
Unit Size:
100 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
16089326 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16089326 Supplier Abnova Supplier No. PAB11259.100ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against synthetic peptide of SLIT2.

Specifications

Antígeno SLIT2
Aplicaciones ELISA, Western Blot
Clasificación Polyclonal
Conjugado Unconjugated
Descripción Rabbit polyclonal antibody raised against synthetic peptide of SLIT2.
Dilución ELISA (1:1500-1:6000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user.
Formulación In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide)
génica SLIT2
Alias de gen FLJ14420/SLIL3/Slit-2
Símbolos de los genes SLIT2
Especie del huésped Rabbit
Inmunógeno A synthetic peptide corresponding to amino acids 484-500 of human SLIT2.
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 9353
Especies diana Bovine, Chicken, Frog, Human, Mouse, Rat
Contenido y almacenamiento Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Formulario Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.