Learn More
SKP1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Descripción
This gene encodes a component of SCF complexes, which are composed of this protein, cullin 1, a ring-box protein, and one member of the F-box family of proteins. This protein binds directly to the F-box motif found in F-box proteins. SCF complexes are involved in the regulated ubiquitination of specific protein substrates, which targets them for degradation by the proteosome. Specific F-box proteins recognize different target protein(s), and many specific SCF substrates have been identified including regulators of cell cycle progression and development. Studies have also characterized the protein as an RNA polymerase II elongation factor. Alternative splicing of this gene results in two transcript variants. A related pseudogene has been identified on chromosome 7. [provided by RefSeq]
Especificaciones
Especificaciones
| Antígeno | SKP1 |
| Aplicaciones | ELISA, Immunohistochemistry, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Descripción | Rabbit polyclonal antibody raised against synthetic peptide of SKP1. |
| Dilución | ELISA (1:2000-1:10000) Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. |
| Formulación | In antiserum (0.01% sodium azide) |
| génica | SKP1 |
| Alias de gen | EMC19/MGC34403/OCP-II/OCP2/SKP1A/TCEB1L/p19A |
| Símbolos de los genes | SKP1 |
| Mostrar más |
For Research Use Only
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.