Learn More
SCRT1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant SCRT1.
Marca: Abnova PAB24402.100uL
Descripción
This gene is a member of the Snail family of C2H2-type zinc finger transcription factors. It codes for a neural-specific transcriptional repressor that binds to E-box motifs. The protein may promote neural differention and may be involved in cancers with neuroendocrine features. [provided by RefSeq
Sequence: LHDKGYLSDYVGPSSVYDGDAEAALLKGPSPEPMYAAAVRGELGPEspecificaciones
| SCRT1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant SCRT1. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| DKFZp547F072/SCRT | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 83482 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) The optimal working dilution should be determined by the end user. | |
| SCRT1 | |
| SCRT1 | |
| Recombinant protein corresponding to amino acids of human SCRT1. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only