Learn More
SCARB2 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of SCARB2.
Marca: Abnova PAB13385.100ug
Descripción
The protein encoded by this gene is a type III glycoprotein that is located primarily in limiting membranes of lysosomes and endosomes. Studies of the similar protein in mice and rat suggested that this protein may participate in membrane transportation and the reorganization of endosomal/lysosomal compartment. Deficiency of the similar protein in mice was reported to impair cell membrane transport processes and cause pelvic junction obstruction, deafness, and peripheral neuropathy. [provided by RefSeq
Especificaciones
| SCARB2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of SCARB2. | |
| In PBS (0.02% sodium azide) | |
| AMRF/CD36L2/HLGP85/LIMPII/SR-BII | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (2-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| SCARB2 | |
| SCARB2 | |
| A synthetic peptide corresponding to C-terminus 18 amino acids of human SCARB2. | |
| RUO | |
| 950 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only