Learn More
S1PR5 Rabbit anti-Human, Monkey, Pig, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of S1PR5.
Marca: Abnova PAB26007.50ug
Descripción
The lysosphingolipid sphingosine 1-phosphate (S1P) regulates cell proliferation, apoptosis, motility, and neurite retraction. Its actions may be both intracellular as a second messenger and extracellular as a receptor ligand. S1P and the structurally related lysolipid mediator lysophosphatidic acid (LPA) signal cells through a set of G protein-coupled receptors known as EDG receptors. Some EDG receptors (e.g., EDG1; MIM 601974) are S1P receptors; others (e.g., EDG2; MIM 602282) are LPA receptors.[supplied by OMIM
Especificaciones
| S1PR5 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of S1PR5. | |
| In PBS (0.09% sodium azide) | |
| EDG8/Edg-8/S1P5/SPPR-1/SPPR-2 | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 53637 | |
| Human, Monkey, Pig, Rat | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (3-6 ug/mL) The optimal working dilution should be determined by the end user. | |
| S1PR5 | |
| S1PR5 | |
| A synthetic peptide corresponding to 17 amino acids at N-terminus of human S1PR5. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only