missing translation for 'onlineSavingsMsg'
Learn More

RYR2, Rabbit, Polyclonal Antibody, Abnova™

Código de producto. 16032727
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
Código de producto. Cantidad unitSize
16032727 100 μg 100 microgramos
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 16032727

Marca: Abnova PAB17291.100ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit polyclonal antibody raised against synthetic peptide of RYR2.

This gene encodes a ryanodine receptor found in cardiac muscle sarcoplasmic reticulum. The encoded protein is one of the components of a calcium channel, composed of a tetramer of the ryanodine receptor proteins and a tetramer of FK506 binding protein 1B proteins, that supplies calcium to cardiac muscle. Mutations in this gene are associated with stress-induced polymorphic ventricular tachycardia and arrhythmogenic right ventricular dysplasia. [provided by RefSeq

Especificaciones

Antígeno RYR2
Aplicaciones Immunofluorescence, Immunohistochemistry (PFA fixed)
Clasificación Polyclonal
Conjugado Unconjugated
Descripción Rabbit polyclonal antibody raised against synthetic peptide of RYR2.
Dilución Immunohistochemistry (1:50-1:100) Immunofluorescence (1:500-1:1000) ELISA (1:5000) The optimal working dilution should be determined by the end user.
Formulación In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol)
génica RYR2
Alias de gen ARVC2/ARVD2/VTSIP
Símbolos de los genes RYR2
Especie del huésped Rabbit
Inmunógeno A synthetic peptide corresponding to internal of human RYR2.
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 6262
Especificidad de la prueba This antibody detects endogenous levels of total RYR2 protein.
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Formulario Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.