Learn More
RELA (phospho S536), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Descripción
NFKB1 (MIM 164011) or NFKB2 (MIM 164012) is bound to REL (MIM 164910), RELA, or RELB (MIM 604758) to form the NFKB complex. The p50 (NFKB1)/p65 (RELA) heterodimer is the most abundant form of NFKB. The NFKB complex is inhibited by I-kappa-B proteins (NFKBIA, MIM 164008 or NFKBIB, MIM 604495), which inactivate NFKB by trapping it in the cytoplasm. Phosphorylation of serine residues on the I-kappa-B proteins by kinases (IKBKA, MIM 600664, or IKBKB, MIM 603258) marks them for destruction via the ubiquitination pathway, thereby allowing activation of the NFKB complex. Activated NFKB complex translocates into the nucleus and binds DNA at kappa-B-binding motifs such as 5-prime GGGRNNYYCC 3-prime or 5-prime HGGARNYYCC 3-prime (where H is A, C, or T; R is an A or G purine; and Y is a C or T pyrimidine).[supplied by OMIM
Especificaciones
Especificaciones
| Antígeno | RELA |
| Aplicaciones | ELISA, Immunohistochemistry (PFA fixed), Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Descripción | Rabbit polyclonal antibody raised against synthetic phosphopeptide of RELA. |
| Dilución | ELISA (1:1000-1:6000) Western Blot (1:200-1:2000) Immunohistochemistry (5 ug/mL) The optimal working dilution should be determined by the end user. |
| Formulación | In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) |
| génica | RELA |
| Alias de gen | MGC131774/NFKB3/p65 |
| Símbolos de los genes | RELA |
| Mostrar más |
For Research Use Only
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.