Learn More
PTH Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant PTH.
Marca: Abnova PAB5103.150ug
Descripción
The protein encoded by this gene is a hormone secreted by parathyroid cells. This hormone elevates blood Ca2+ level by dissolving the salts in bone and preventing their renal excretion. Defects in this gene are a cause of familial isolated hypoparathyroidism (FIH). [provided by RefSeq]
Especificaciones
| PTH | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant PTH. | |
| In PBS, pH 7.5 (50% glycerol, 0.01% thimerosal) | |
| PTH1 | |
| Rabbit | |
| 150 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Immunoprecipitation | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| PTH | |
| PTH | |
| Recombinant protein corresponding to human PTH. | |
| RUO | |
| 5741 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only