Learn More
PRKCB Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PRKCB.
Marca: Abnova PAB4532.400uL
Descripción
Protein kinase C (PKC) is a family of serine- and threonine-specific protein kinases that can be activated by calcium and second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC family members also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play a distinct role in cells. The protein encoded by this gene is one of the PKC family members. This protein kinase has been reported to be involved in many different cellular functions, such as B cell activation, apoptosis induction, endothelial cell proliferation, and intestinal sugar absorption. Studies in mice also suggest that this kinase may also regulate neuronal functions and correlate fear-induced conflict behavior after stress. Alternatively spliced transcript variants encoding distinct isoforms have been reported. [provided by RefSeq
Especificaciones
| PRKCB | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of PRKCB. | |
| In PBS (0.09% sodium azide) | |
| MGC41878/PKC-beta/PKCB/PRKCB1/PRKCB2 | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 5579 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| ELISA (1:1000) Western Blot (1:100-500) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
| PRKCB | |
| PRKCB | |
| A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human PRKCB. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only