Learn More
PCTK2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of PCTK2.
Marca: Abnova PAB2947.400uL
Descripción
The protein encoded by this gene belongs to the cdc2/cdkx subfamily of the ser/thr family of protein kinases. It has similarity to rat protein which is thought to play a role in terminally differentiated neurons. [provided by RefSeq
Especificaciones
| PCTK2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of PCTK2. | |
| In PBS (0.09% sodium azide) | |
| PCTAIRE2 | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 5128 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
| PCTK2 | |
| PCTK2 | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human PCTK2. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only