Learn More
NXF3 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Descripción
This gene is one member of a family of nuclear RNA export factor genes. Common domain features of this family are a noncanonical RNP-type RNA-binding domain (RBD), 4 leucine-rich repeats (LRRs), a nuclear transport factor 2 (NTF2)-like domain that allows heterodimerization with NTF2-related export protein-1 (NXT1), and a ubiquitin-associated domain that mediates interactions with nucleoporins. The LRRs and NTF2-like domains are required for export activity. Alternative splicing seems to be a common mechanism in this gene family. The encoded protein of this gene has shortened LRR and ubiquitin-associated domains and its RDB is unable to bind RNA. It is located in the nucleoplasm but is not associated with either the nuclear envelope or the nucleolus. [provided by RefSeq
Especificaciones
Especificaciones
| Antígeno | NXF3 |
| Aplicaciones | Immunohistochemistry |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Descripción | Rabbit polyclonal antibody raised against recombinant NXF3. |
| Dilución | Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. |
| Formulación | In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide). |
| génica | NXF3 |
| Símbolos de los genes | NXF3 |
| Especie del huésped | Rabbit |
| Mostrar más |
For Research Use Only
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.