missing translation for 'onlineSavingsMsg'
Learn More

NUP155 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Código de producto. 16040387
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
Código de producto. Cantidad unitSize
16040387 100 μg 100 microgramos
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 16040387

Marca: Abnova PAB13051.100ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit polyclonal antibody raised against synthetic peptide of NUP155.

Nucleoporins are the main components of the nuclear pore complex (NPC) of eukaryotic cells. They are involved in the bidirectional trafficking of molecules, especially mRNAs and proteins, between the nucleus and the cytoplasm. The protein encoded by this gene does not contain the typical FG repeat sequences found in most vertebrate nucleoporins. Two protein isoforms are encoded by transcript variants of this gene. [provided by RefSeq

Especificaciones

Antígeno NUP155
Aplicaciones Western Blot
Clasificación Polyclonal
Conjugado Unconjugated
Descripción Rabbit polyclonal antibody raised against synthetic peptide of NUP155.
Dilución Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Formulación In PBS (0.02% sodium azide)
génica NUP155
Alias de gen KIAA0791/N155
Símbolos de los genes NUP155
Especie del huésped Rabbit
Inmunógeno A synthetic peptide corresponding to C-terminus 18 amino acids of human NUP155.
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 9631
Especies diana Human, Mouse
Contenido y almacenamiento Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Formulario Liquid
Mostrar más Mostrar menos

For Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.