Learn More
Invitrogen™ Rabbit anti-Mouse IgG (H+L) Secondary Antibody, Rhodamine
Descripción
Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31665 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31665 reacts with the heavy chains of mouse IgG and with the light chains common to most mouse immunoglobulins, but does not react against non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulin from other species. Store product protected from light at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. Rhodamine Amax= 550 nm; Emax= 570 nm. Fluorophore/Protein Absorbance Ratio: A550/A280 = ~ 0.4 (lot-dependent). Reconstitute with 1.1 mL of distilled water (1.5 mg/mL after restoration). Country of Origin: USA.
Especificaciones
Especificaciones
| Antígeno | Mouse IgG (H+L) |
| Aplicaciones | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry |
| Clasificación | Polyclonal |
| Concentración | 1.5 mg/mL |
| Conjugado | Rhodamine |
| Formulación | PBS with 15mg/mL BSA and 0.05% sodium azide; pH 7.6 |
| Especie del huésped | Rabbit |
| Método de purificación | Affinity chromatography |
| Cantidad | 1.5 mg |
| Estado normativo | RUO |
| Mostrar más |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.