Learn More
MAP1LC3A Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of MAP1LC3A.
Marca: Abnova PAB12389.100uL
Descripción
MAP1A and MAP1B are microtubule-associated proteins which mediate the physical interactions between microtubules and components of the cytoskeleton. MAP1A and MAP1B each consist of a heavy chain subunit and multiple light chain subunits. The protein encoded by this gene is one of the light chain subunits and can associate with either MAP1A or MAP1B. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Especificaciones
| MAP1LC3A | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of MAP1LC3A. | |
| In PBS (30% glycerol, 0.09% sodium azide) | |
| LC3/LC3A/MAP1ALC3/MAP1BLC3 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| This antibody is specific to LC3. | |
| Store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200-1:400) Immunoprecipitation (20 ug/500 ug of protein) Western Blot (2 ug/mL) The optimal working dilution should be determined by the end user. | |
| MAP1LC3A | |
| MAP1LC3A | |
| A synthetic peptide corresponding to amino acids 1-100 of human MAP1LC3A. | |
| RUO | |
| 84557 | |
| Human, Mouse, Rat | |
| Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only