Learn More
LPHN2 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Código de producto. 16096127
Change view
Click to view available options
Cantidad:
50 μg
Tamaño de la unidad:
50 microgramos
16096127 50 μg 50 microgramos
1 options
Código de producto. 16096127
Marca: Abnova PAB25949.50ug
Rabbit polyclonal antibody raised against synthetic peptide of LPHN2.
This gene encodes a member of the latrophilin subfamily of G-protein coupled receptors (GPCR). Latrophilins may function in both cell adhesion and signal transduction. In experiments with nonhuman species, endogenous proteolytic cleavage within a cysteine-rich GPS (G-protein-coupled-receptor proteolysis site) domain resulted in two subunits (a large extracellular N-terminal cell adhesion subunit and a subunit with substantial similarity to the secretin/calcitonin family of GPCRs) being noncovalently bound at the cell membrane. While several transcript variants have been described, the biological validity of only one has been determined. [provided by RefSeq
Especificaciones
Antígeno LPHN2
Aplicaciones Immunohistochemistry (PFA fixed)
Clasificación Polyclonal
Conjugado Unconjugated
Descripción Rabbit polyclonal antibody raised against synthetic peptide of LPHN2.
Dilución Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (18-45 ug/mL) The optimal working dilution should be determined by the end user.
Formulación In PBS (0.09% sodium azide)
génica LPHN2
Alias de gen CIRL2/CL2/LEC1/LPHH1
Símbolos de los genes LPHN2
Especie del huésped Rabbit
Inmunógeno A synthetic peptide corresponding to 16 amino acids at N-terminus of human LPHN2.
Método de purificación Immunoaffinity chromatography
Cantidad 50 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 23266
Especificidad de la prueba BLAST analysis of the peptide immunogen showed no homology with other human proteins.
Especies diana Human
Contenido y almacenamiento Store at 4°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Formulario Liquid
Mostrar más Mostrar menos
For Research Use Only