Learn More
KLK5 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of KLK5.
Marca: Abnova PAB4411.400uL
Descripción
Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. This gene is one of the fifteen kallikrein subfamily members located in a cluster on chromosome 19. Its expression is up-regulated by estrogens and progestins. The encoded protein is secreted and may be involved in desquamation in the epidermis. Alternative splicing results in multiple transcript variants encoding the same protein. [provided by RefSeq
Especificaciones
| KLK5 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of KLK5. | |
| In PBS (0.09% sodium azide) | |
| KLK-L2/KLKL2/SCTE | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| 25818 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:1000) Western Blot (1:100-500) The optimal working dilution should be determined by the end user. | |
| KLK5 | |
| KLK5 | |
| A synthetic peptide (conjugated with KLH) corresponding to internal region of human KLK5. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only