Learn More
KCNJ9, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of KCNJ9.
Marca: Abnova PAB17723.100ug
Descripción
Potassium channels are present in most mammalian cells, where they participate in a wide range of physiologic responses. The protein encoded by this gene is an integral membrane protein and inward-rectifier type potassium channel. The encoded protein, which has a greater tendency to allow potassium to flow into a cell rather than out of a cell, is controlled by G-proteins. It associates with another G-protein-activated potassium channel to form a heteromultimeric pore-forming complex. [provided by RefSeq
Especificaciones
| KCNJ9 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of KCNJ9. | |
| In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
| GIRK3/KIR3.3 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody detects endogenous levels of total KCNJ9 protein. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| Western Blot (1:500-1:1000) ELISA (1:40000) The optimal working dilution should be determined by the end user. | |
| KCNJ9 | |
| KCNJ9 | |
| A synthetic peptide corresponding to internal of human KCNJ9. | |
| RUO | |
| 3765 | |
| Human, Mouse, Rat | |
| Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only