Learn More
ISG15 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ISG15.
Brand: Abnova PAB1719.400uL
Description
ISG15 is a ubiquitin-like protein that becomes conjugated to many cellular proteins upon activation by interferon-alpha (IFNA; MIM 147660) and -beta (IFNB; MIM 147640) (Zhao et al., 2005 [PubMed 16009940]).[supplied by OMIM
Specifications
| ISG15 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ISG15. | |
| In PBS (0.09% sodium azide) | |
| G1P2/IFI15/UCRP | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 9636 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| ISG15 | |
| ISG15 | |
| A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human ISG15. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only