Learn More
IL10 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant IL10.
Marca: Abnova PAB19879.150ug
Descripción
The protein encoded by this gene is a cytokine produced primarily by monocytes and to a lesser extent by lymphocytes. This cytokine has pleiotropic effects in immunoregulation and inflammation. It down-regulates the expression of Th1 cytokines, MHC class II Ags, and costimulatory molecules on macrophages. It also enhances B cell survival, proliferation, and antibody production. This cytokine can block NF-kappa B activity, and is involved in the regulation of the JAK-STAT signaling pathway. Knockout studies in mice suggested the function of this cytokine as an essential immunoregulator in the intestinal tract. [provided by RefSeq
Especificaciones
| IL10 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant IL10. | |
| In PBS, pH 7.5 (0.02% sodium azide, 50% glycerol (v/v)) | |
| CSIF/IL-10/IL10A/MGC126450/MGC126451/TGIF | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 3586 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| IL10 | |
| IL10 | |
| Recombinant protein corresponding to human IL10. | |
| 150 μg | |
| Primary | |
| Human | |
| Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only