missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Horse IgG whole molecule,Polyclonal Secondary Antibody, Abnova™
Marca: Abnova PAB9273.2mL
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
| Rabbit anti-Horse IgG whole molecule | |
| Polyclonal | |
| Rabbit anti-horse IgG recognizes horse IgG whole molecule. This secondary antibody was purified using delipidation and defibrination. | |
| Lyophilized from 20mM KH2PO4, 150mM NaCl, pH 7.2 | |
| Horse IgG whole molecule | |
| 2 mL | |
| Equine | |
| Lyophilized |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| This product was prepared from monospecific antiserum by a delipidation and defibrination. Assay by immunoelectrophoresis resulted in a single precipitin arc against horse IgG and horse serum. | |
| Secondary | |
| Store at 4°C on dry atmosphere. After reconstitution with 2mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido