Learn More
GSK3A (phospho S21), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of GSK3A.
Marca: Abnova PAB12623.100ug
Descripción
Glycogen synthase kinase 3-alpha (GSK3A; EC {2.7.1.37}) is a multifunctional protein serine kinase, homologous to Drosophila 'shaggy' (zeste-white3) and implicated in the control of several regulatory proteins including glycogen synthase (see GYS1, {138570}) and transcription factors (e.g., JUN, {165160}). It also plays a role in the WNT ({164820}) and PI3K (see PIK3CG, {601232}) signaling pathways (see review by {1:Ali et al. (2001)}).[supplied by OMIM
Sequence: RTSSFASpécification
| GSK3A | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of GSK3A. | |
| In TBS, pH 7.2 (BSA, 10% Proclin300) | |
| DKFZp686D0638 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody recognizes GSK3A (pS21) with a phosphorylated site at Serine 9. It does not cross-react with non-phosphospecific peptide. | |
| Store at 4°C. For long term storage store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Western Blot (0.1-1 ug/mL) ELISA (0.01-0.1 ug/mL) Immunoprecipitation (2-5 ug/mL) The optimal working dilution should be determined by the end user. | |
| GSK3A | |
| GSK3A | |
| Synthetic phosphopeptide corresponding to residues surrounding S21 of human GSK3A. | |
| RUO | |
| 2931 | |
| Human, Mouse, Rat | |
| Liquid |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
For Research Use Only