missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR17 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of GPR17.
Marca: Abnova PAB26324.50ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
| GPR17 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of GPR17. | |
| In PBS (0.09% sodium azide) | |
| Q13304 | |
| GPR17 | |
| A synthetic peptide corresponding to 16 amino acids at N-terminal extracellular domain of human GPR17. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Immunocytochemistry, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (3 ug/mL) The optimal working dilution should be determined by the end user. | |
| GPR17 | |
| DKFZp686M18273 | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 2840 | |
| Human | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido