Learn More
Rabbit anti-Golden Syrian hamster serum proteins, Polyclonal, Secondary Antibody, Abnova™
Especificaciones
Especificaciones
| Antígeno | Rabbit anti-Golden Syrian hamster serum proteins |
| Aplicaciones | ELISA, Immunohistochemistry, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Descripción | Rabbit anti-Golden Syrian hamster IgG recognizes Golden Syrian hamster serum proteins. This secondary antibody was purified using delipidation and defibrination. |
| Dilución | The optimal working dilution should be determined by the end user. |
| Formulación | Lyophilized from 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) |
| Especie del huésped | Rabbit |
| Inmunógeno | Golden Syrian hamster serum proteins |
| Método de purificación | This product was prepared from polyspecific antiserum by a delipidationand defibrination. Assay by immunoelectrophoresis resulted in a multipleprecipitin arcs against Golden Syrian hamster serum. |
| Mostrar más |
For Research Use Only