Learn More
FTO, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of FTO.
Marca: Abnova PAB16736.100ug
Description
The exact function of this gene is not know. Studies in mice suggest that it may be involved in nucleic acid demethylation, and that its mRNA level is regulated by feeding and fasting. Genomewide association studies of type 2 diabetes indicate this gene as a diabetes susceptibility locus. Mutation in this gene has been associated with growth retardation, developmental delay, coarse facies, and early death. [provided by RefSeq
Specifications
| FTO | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of FTO. | |
| In PBS (0.02% sodium azide) | |
| Q9C0B1 | |
| FTO | |
| A synthetic peptide corresponding to N-terminus 15 amino acids of human FTO. | |
| RUO | |
| 79068 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| FTO | |
| KIAA1752/MGC5149 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only