missing translation for 'onlineSavingsMsg'
Learn More

Fkbp1a, Rabbit, Polyclonal Antibody, Abnova™

Código de producto. 16021198
Change view
Click to view available options
Cantidad:
100 μg
Tamaño de la unidad:
100 microgramos
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
16021198 100 μg 100 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16021198 Supplier Abnova Supplier No. PAB8720.100ug

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
View alternative products

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against synthetic peptide of Fkbp1a.

Specifications

Antígeno Fkbp1a
Aplicaciones Immunohistochemistry, Immunoprecipitation, Western Blot
Clasificación Polyclonal
Conjugado Unconjugated
Descripción Rabbit polyclonal antibody raised against synthetic peptide of Fkbp1a.
Dilución Western Blot (0.5-4 ug/mL) Immunoprecipitation (2-10 ug/mL) Immunohistochemistry (2-10 ug/mL) The optimal working dilution should be determined by the end user.
Formulación In PBS, pH 7.2 (30% glycerol, 0.5mg/mL BSA, 0.01% thimerosal)
génica Fkbp1a
Alias de gen 12kDa/FKBP12/FKBP12-T1/FKBP12-T2/Fkbp/Fkbp1/mFKBP1/mFKBP12
Símbolos de los genes Fkbp1a
Especie del huésped Rabbit
Inmunógeno A synthetic peptide corresponding to residues surrounding amino acids 10 of mouse Fkbp1a.
Cantidad 100 μg
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 14225
Especificidad de la prueba This antibody detects 12 KDa FKBP12.
Especies diana Bovine, Human, Mouse, Rabbit, Rat
Contenido y almacenamiento Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Formulario Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.