missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Esr1 Rabbit anti-Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide Esr1.
Marca: Abnova PAB0115.50uL
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
| Esr1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide Esr1. | |
| Esr1 | |
| AA420328/AU041214/ER-alpha/ER[a]/ERa/ERalpha/ESR/Estr/Estra/Nr3a1 | |
| Rabbit | |
| 50 μL | |
| Primary | |
| This antibody reacts very weakly with ER-alpha (67 KDa band), but recognizes ER-X (62 KDa band) very well in the mouse brain and uterus. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
| NM_007956.3 | |
| Esr1 | |
| A synthetic peptide from the C-terminus end of the mouse estrogen receptor alpha. | |
| RUO | |
| 13982 | |
| Mouse | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido