Learn More
DKK4 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of DKK4.
Marca: Abnova PAB3573.400uL
Descripción
This gene encodes a protein that is a member of the dickkopf family. The secreted protein contains two cysteine rich regions and is involved in embryonic development through its interactions with the Wnt signaling pathway. Activity of this protein is modulated by binding to the Wnt co-receptor and the co-factor kremen 2. [provided by RefSeq
Especificaciones
| DKK4 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of DKK4. | |
| In PBS (0.09% sodium azide) | |
| NP_055235 | |
| DKK4 | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human DKK4. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-100) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| DKK4 | |
| DKK-4/MGC129562/MGC129563 | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 27121 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only