Learn More
DDX3X Rabbit anti-Human, Polyclonal Antibody, Abnova™
Descripción
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein, which interacts specifically with hepatitis C virus core protein resulting a change in intracellular location. This gene has a homolog located in the nonrecombining region of the Y chromosome. The protein sequence is 91% identical between this gene and the Y-linked homolog. [provided by RefSeq
Especificaciones
Especificaciones
| Antígeno | DDX3X |
| Aplicaciones | Immunohistochemistry (PFA fixed), Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Descripción | Rabbit polyclonal antibody raised against synthetic peptide of DDX3X. |
| Dilución | Western Blot (1:1000) Immunohistochemistry (1:10-50) The optimal working dilution should be determined by the end user. |
| Formulación | In PBS (0.09% sodium azide) |
| génica | DDX3X |
| N.º de referencia del gen | NP_001347;O00571 |
| Alias de gen | DBX/DDX14/DDX3/HLP2 |
| Mostrar más |
For Research Use Only
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.