Learn More
CPSF3L Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant CPSF3L.
Marca: Abnova PAB22238.100uL
Descripción
The Integrator complex contains at least 12 subunits and associates with the C-terminal domain of RNA polymerase II large subunit (POLR2A; MIM 180660) and mediates the 3-prime end processing of small nuclear RNAs U1 (RNU1; MIM 180680) and U2 (RNU2; MIM 180690). INTS11, or CPSF3L, is the catalytic subunit of the Integrator complex (Baillat et al., 2005 [PubMed 16239144]).[supplied by OMIM
Sequence: LVGQAEPESVLLVHGEAKKMEFLKQKIEQELRVNCYMPANGETVTLPTSPSIPVGISLGLLKREMAQGLLPEAKKPRLLHGTLIMKDSNFRLVSSEQALKEspecificaciones
| CPSF3L | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant CPSF3L. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q5TA45 | |
| CPSF3L | |
| Recombinant protein corresponding to amino acids of human CPSF3L. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| CPSF3L | |
| CPSF73L/FLJ13294/FLJ20542/INT11/INTS11/RC-68/RC68 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 54973 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only