missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CITED2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CITED2.
Marca: Abnova PAB25612.100ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
| CITED2 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| CITED2 | |
| MRG1/P35SRJ | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 10370 | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of CITED2. | |
| In PBS (0.05% sodium azide) | |
| NP_001161861 | |
| CITED2 | |
| A synthetic peptide corresponding to 18 amino acids at N-terminus of human CITED2. | |
| 100 μg | |
| Primary | |
| Multiple isoforms of CITED2 are known to exist. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido