Learn More
CASC3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of CASC3.
Marca: Abnova PAB3061.400uL
Descripción
The product of this gene is a core component of the exon junction complex (EJC), a protein complex that is deposited on spliced mRNAs at exon-exon junctions and functions in nonsense-mediated mRNA decay (NMD). The encoded protein binds RNA and interacts with two other EJC core components. It is predominantly located in the cytoplasm, but shuttles into the nucleus where it localizes to nuclear speckles. [provided by RefSeq
Especificaciones
| CASC3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of CASC3. | |
| In PBS (0.09% sodium azide) | |
| Q8K3W3;NP_619601 | |
| CASC3 | |
| Synthetic phosphopeptide (conjugated with KLH) corresponding to amino acids 166-196 residues surrounding Y181 of human CASC3. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) Immunohistochemistry (1:10-50) The optimal working dilution should be determined by the end user. | |
| CASC3 | |
| BTZ/MLN51 | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 22794 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only