Learn More
BLNK Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Descripción
This gene encodes a cytoplasmic linker or adaptor protein that plays a critical role in B cell development. This protein bridges B cell receptor-associated kinase activation with downstream signaling pathways, thereby affecting various biological functions. The phosphorylation of five tyrosine residues is necessary for this protein to nucleate distinct signaling effectors following B cell receptor activation. Mutations in this gene cause hypoglobulinemia and absent B cells, a disease in which the pro- to pre-B-cell transition is developmentally blocked. Deficiency in this protein has also been shown in some cases of pre-B acute lymphoblastic leukemia. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Especificaciones
Especificaciones
| Antígeno | BLNK |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Descripción | Rabbit polyclonal antibody raised against partial recombinant BLNK. |
| Dilución | The optimal working dilution should be determined by the end user. |
| Formulación | In PBS, pH 7.4 (0.09% sodium azide) |
| génica | BLNK |
| Alias de gen | BASH/BLNK-S/LY57/MGC111051/SLP-65/SLP65 |
| Símbolos de los genes | BLNK |
| Mostrar más |
For Research Use Only
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.