Learn More
BDNF Rabbit anti-Human, Rat, Polyclonal Antibody, Abnova™
Descripción
The protein encoded by this gene is a member of the nerve growth factor family. It is induced by cortical neurons, and is necessary for survival of striatal neurons in the brain. Expression of this gene is reduced in both Alzheimer's and Huntington disease patients. This gene may play a role in the regulation of stress response and in the biology of mood disorders. Multiple transcript variants encoding distinct isoforms have been described for this gene. [provided by RefSeq]
Especificaciones
Especificaciones
| Antígeno | BDNF |
| Aplicaciones | ELISA, Immunohistochemistry (Frozen), Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Descripción | Rabbit polyclonal antibody raised against synthetic peptide of BDNF. |
| Dilución | Immunohistochemistry (1:1000-1:2000) ELISA (1:1000-1:2000) Western Blot (1:1000-1:2000) The optimal working dilution should be determined by the end user. |
| Formulación | Lyophilized from PBS |
| génica | BDNF |
| N.º de referencia del gen | P23560 |
| Alias de gen | MGC34632 |
| Mostrar más |
For Research Use Only
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.