missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATF6 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ATF6.
Marca: Abnova PAB13180.100ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
ATF6 is an endoplasmic reticulum (ER) stress-regulated transmembrane transcription factor that activates the transcription of ER molecules.[supplied by OMIM
Especificaciones
| ATF6 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ATF6. | |
| In PBS (0.02% sodium azide) | |
| NP_031374 | |
| ATF6 | |
| A synthetic peptide corresponding to N-terminus 17 amino acids of human ATF6. | |
| RUO | |
| 22926 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunocytochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| ATF6 | |
| ATF6A | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido