Learn More
ARFGAP3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant ARFGAP3.
Marca: Abnova PAB19985.100uL
Descripción
The protein encoded by this gene is a GTPase-activating protein (GAP) that associates with the Golgi apparatus and regulates the early secretory pathway of proteins. The encoded protein promotes hydrolysis of ADP-ribosylation factor 1 (ARF1)-bound GTP, which is required for the dissociation of coat proteins from Golgi-derived membranes and vesicles. Dissociation of the coat proteins is a prerequisite for the fusion of these vesicles with target compartments. The activity of this protein is sensitive to phospholipids. Multiple transcript variants encoding different isoforms have been found for this gene. This gene was originally known as ARFGAP1, but that is now the name of a related but different gene. [provided by RefSeq
Sequence: SKQDILTIFKRLRSVPTNKVCFDCGAKNPSWASITYGVFLCIDCSGSHRSLGVHLSFIRSTELDSNWSWFQLRCMQVGGNASASSFFHQHGCSTNDTNAKYNSRAAQLYREKIKSLASQATRKHGTDLWLDSCVVPPLSPPPEspecificaciones
| ARFGAP3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant ARFGAP3. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q9NP61 | |
| ARFGAP3 | |
| Recombinant protein corresponding to amino acids of human ARFGAP3. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| ARFGAP3 | |
| ARFGAP1/FLJ45618 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 26286 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only