Learn More
AKT3 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Descripción
The protein encoded by this gene is a member of the AKT, also called PKB, serine/threonine protein kinase family. AKT kinases are known to be regulators of cell signaling in response to insulin and growth factors. They are involved in a wide variety of biological processes including cell proliferation, differentiation, apoptosis, tumorigenesis, as well as glycogen synthesis and glucose uptake. This kinase has been shown to be stimulated by platelet-derived growth factor (PDGF), insulin, and insulin-like growth factor 1 (IGF1). Alternatively splice transcript variants encoding distinct isoforms have been described. [provided by RefSeq
Especificaciones
Especificaciones
| Antígeno | AKT3 |
| Aplicaciones | ELISA, Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Descripción | Rabbit polyclonal antibody raised against synthetic peptide of AKT3. |
| Dilución | ELISA (1:1000) Western Blot (1:50-100) The optimal working dilution should be determined by the end user. |
| Formulación | In PBS (0.09% sodium azide) |
| génica | AKT3 |
| N.º de referencia del gen | NP_005456;Q9Y243 |
| Alias de gen | DKFZp434N0250/PKB-GAMMA/PKBG/PRKBG/RAC-PK-gamma/RAC-gamma/STK-2 |
| Mostrar más |
For Research Use Only
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.