Learn More
ADAM17 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ADAM17.
Marca: Abnova PAB1888.400uL
Descripción
This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biologic processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. The protein encoded by this gene functions as a tumor necrosis factor-alpha converting enzyme; binds mitotic arrest deficient 2 protein; and also plays a prominent role in the activation of the Notch signaling pathway. [provided by RefSeq
Especificaciones
| ADAM17 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of ADAM17. | |
| In PBS (0.09% sodium azide) | |
| NP_003174;P78536 | |
| ADAM17 | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human ADAM17. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Flow Cytometry, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) Flow cytometry (1:10-50) The optimal working dilution should be determined by the end user. | |
| ADAM17 | |
| CD156b/MGC71942/TACE/cSVP | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| 6868 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
For Research Use Only