Learn More
ACE Rabbit anti-Human, Polyclonal Antibody, Abnova™
Descripción
This gene encodes an enzyme involved in catalyzing the conversion of angiotensin I into a physiologically active peptide angiotensin II. Angiotensin II is a potent vasopressor and aldosterone-stimulating peptide that controls blood pressure and fluid-electrolyte balance. This enzyme plays a key role in the renin-angiotensin system. Many studies have associated the presence or absence of a 287bp Alu repeat element in this gene with the levels of circulating enzyme or cardiovascular pathophysiologies. Two most abundant alternatively spliced variants of this gene encode two isozymes - the somatic form and the testicular form that are equally active. Multiple additional alternatively spliced variants have been identified but their full length nature has not been determined. [provided by RefSeq
Especificaciones
Especificaciones
| Antígeno | ACE |
| Aplicaciones | Immunohistochemistry (PFA fixed) |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Descripción | Rabbit polyclonal antibody raised against recombinant ACE. |
| Dilución | Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. |
| Formulación | In PBS, pH 7.2, (40% glycerol, 0.02% sodium azide) |
| génica | ACE |
| Alias de gen | ACE1/CD143/DCP/DCP1/MGC26566 |
| Símbolos de los genes | ACE |
| Mostrar más |
For Research Use Only
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.